Assay Details
| Gene Symbol: CCDC113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257353.1 | XP_008255575.1 | 3 - 4 | 479 | 71 | |
| XM_008257354.1 | XP_008255576.1 | 3 - 4 | 479 | 71 | ||
| XM_008257356.2 | XP_008255578.1 | 3 - 4 | 479 | 71 | ||
More Information
Set Membership: |
|

CCDC113
Genomic Map