Assay Details
| Gene Symbol: SRPX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720389.3 | XP_002720435.1 | 11 - 12 | 1687 | 58 | |
| XM_008273146.2 | XP_008271368.1 | 11 - 12 | 1814 | 58 | ||
More Information
Set Membership: |
|

SRPX2
Genomic Map