Assay Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709681.3 | XP_002709727.2 | 2 - 3 | 874 | 110 | |
| XM_008254183.2 | XP_008252405.1 | 2 - 3 | 882 | 110 | ||
| XM_008254184.2 | XP_008252406.1 | 2 - 3 | 966 | 110 | ||
| XM_008254185.2 | XP_008252407.1 | 2 - 3 | 877 | 110 | ||
| XM_008254186.2 | XP_008252408.1 | 2 - 3 | 967 | 110 | ||
| XM_017340434.1 | XP_017195923.1 | 2 - 3 | 663 | 110 | ||
| XM_017340438.1 | XP_017195927.1 | 2 - 3 | 966 | 110 | ||
| XM_017340441.1 | XP_017195930.1 | 2 - 3 | 965 | 110 | ||
| XM_017340443.1 | XP_017195932.1 | 2 - 3 | 964 | 110 | ||
More Information
Set Membership: |
|

TRABD2A
Genomic Map