Assay Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709681.3 | XP_002709727.2 | 5 - 6 | 1283 | 78 | |
| XM_008254184.2 | XP_008252406.1 | 5 - 6 | 1339 | 78 | ||
| XM_017340434.1 | XP_017195923.1 | 5 - 6 | 1072 | 78 | ||
| XM_017340438.1 | XP_017195927.1 | 5 - 6 | 1386 | 78 | ||
More Information
Set Membership: |
|

TRABD2A
Genomic Map