Assay Details
| Gene Symbol: DHX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721247.3 | XP_002721293.1 | 2707 | 52 | ||
| XM_017350760.1 | XP_017206249.1 | 2778 | 52 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHX32| Gene Symbol: DHX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721247.3 | XP_002721293.1 | 2707 | 52 | ||
| XM_017350760.1 | XP_017206249.1 | 2778 | 52 | |||
Set Membership: |
|