Assay Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711268.3 | XP_002711314.1 | 3 - 4 | 617 | 93 | |
| XM_008256852.2 | XP_008255074.1 | 3 - 4 | 515 | 93 | ||
More Information
Set Membership: |
|

CCDC59
Genomic Map