Assay Details
| Gene Symbol: GLDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717725.3 | XP_002717771.2 | 5 - 6 | 1003 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLDN| Gene Symbol: GLDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717725.3 | XP_002717771.2 | 5 - 6 | 1003 | 59 | |
Set Membership: |
|