Assay Details
| Gene Symbol: PRELID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710425.3 | XP_002710471.1 | 2 - 3 | 459 | 82 | |
| XM_017341275.1 | XP_017196764.1 | 2 - 3 | 459 | 82 | ||
More Information
Set Membership: |
|

PRELID1
Genomic Map