Assay Details
Gene Symbol: LOC4350758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761536.1 | XP_015617022.1 | 12 - 13 | 1870 | 72 | |
GenBank mRNA | EF417892.1 | - | 12 - 13 | 1326 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
