Assay Details
Gene Symbol: LOC4350758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761536.1 | XP_015617022.1 | 8 - 9 | 1399 | 70 | |
GenBank mRNA | EF417892.1 | - | 8 - 9 | 855 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
