Assay Details
| Gene Symbol: LOC4337651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782106.2 | XP_015637592.1 | 2 - 3 | 449 | 90 | |
| XM_015782107.2 | XP_015637593.1 | 2 - 3 | 449 | 90 | ||
| GenBank mRNA | AK066388.1 | - | 2 - 3 | 407 | 90 | |
| AK120088.1 | - | 2 - 3 | 405 | 90 | ||
More Information
Set Membership: |
|

LOC4337651
Genomic Map