Assay Details
Gene Symbol: LOC4339406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782182.2 | XP_015637668.1 | 2 - 3 | 500 | 58 | |
XM_015782183.2 | XP_015637669.1 | 2 - 3 | 497 | 58 | ||
GenBank mRNA | AK062298.1 | - | 2 - 3 | 459 | 58 | |
AK064168.1 | - | 2 - 3 | 458 | 58 | ||
AK108715.1 | - | 2 - 3 | 453 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
