Assay Details
Gene Symbol: LOC4330324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767985.2 | XP_015623471.1 | 1 | 880 | 83 | |
GenBank mRNA | AK071853.1 | - | 1 | 853 | 83 | |
AK099314.1 | - | 1 | 853 | 83 | ||
AK104122.1 | - | 1 | 847 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
