Assay Details
Gene Symbol: LOC4334172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777686.2 | 4 - 5 | 1406 | 127 | ||
GenBank mRNA | AK060339.1 | - | 2 - 3 | 410 | 127 | |
AK067626.1 | - | 5 - 6 | 1157 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
