Assay Details
| Gene Symbol: LOC4334408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058269.1 | - | 2 - 3 | 180 | 72 | |
| AK070933.1 | - | 19 - 20 | 2210 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4334408| Gene Symbol: LOC4334408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058269.1 | - | 2 - 3 | 180 | 72 | |
| AK070933.1 | - | 19 - 20 | 2210 | 72 | ||
Set Membership: |
|