Assay Details
| Gene Symbol: LOC4334408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058269.1 | - | 4 - 5 | 331 | 81 | |
| AK070933.1 | - | 21 - 22 | 2361 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4334408| Gene Symbol: LOC4334408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058269.1 | - | 4 - 5 | 331 | 81 | |
| AK070933.1 | - | 21 - 22 | 2361 | 81 | ||
Set Membership: |
|