Assay Details
Gene Symbol: LOC4342724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789737.2 | XP_015645223.1 | 1 | 584 | 114 | |
GenBank mRNA | AK242298.1 | - | 1 | 576 | 114 | |
D42141.1 | - | 1 | 555 | 115 |
Gene Symbol: LOC4334019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK288076.1 | - | 577 | 114 |
Gene Symbol: LOC4334201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK287934.1 | - | 1435 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
