Assay Details
| Gene Symbol: LOC4345636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795144.2 | XP_015650630.1 | 5 - 6 | 647 | 95 | |
| GenBank mRNA | AK102785.1 | - | 5 - 6 | 603 | 95 | |
More Information
Set Membership: |
|

LOC4345636
Genomic Map