Assay Details
| Gene Symbol: LOC4327315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766628.2 | XP_015622114.1 | 2 - 3 | 1388 | 69 | |
| GenBank mRNA | AK120715.1 | - | 2 - 3 | 1394 | 69 | |
| AY323485.1 | - | 2 - 3 | 1369 | 69 | ||
| DQ450947.1 | - | 2 - 3 | 1330 | 69 | ||
| HM991169.1 | - | 2 - 3 | 1305 | 69 | ||
More Information
Set Membership: |
|

LOC4327315
Genomic Map