Assay Details
| Gene Symbol: LOC4327315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766628.2 | XP_015622114.1 | 3 - 4 | 1595 | 62 | |
| XM_015766629.2 | XP_015622115.1 | 3 - 4 | 1608 | 62 | ||
| GenBank mRNA | AK120715.1 | - | 3 - 4 | 1601 | 62 | |
| AY323485.1 | - | 3 - 4 | 1576 | 62 | ||
| DQ450947.1 | - | 3 - 4 | 1537 | 62 | ||
| HM991169.1 | - | 3 - 4 | 1512 | 62 | ||
More Information
Set Membership: |
|

LOC4327315
Genomic Map