Assay Details
| Gene Symbol: LOC4324662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766908.2 | XP_015622394.1 | 1 - 2 | 294 | 148 | |
| XM_015766912.2 | XP_015622398.1 | 1 - 2 | 294 | 148 | ||
| GenBank mRNA | AK066657.1 | - | 1 - 2 | 101 | 148 | |
More Information
Set Membership: |
|

LOC4324662
Genomic Map