Assay Details
| Gene Symbol: LOC4350807 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761490.2 | XP_015616976.1 | 14 - 15 | 3402 | 74 | |
| GenBank mRNA | AK121517.1 | - | 11 - 12 | 3716 | 74 | |
More Information
Set Membership: |
|

LOC4350807
Genomic Map