Assay Details
| Gene Symbol: LOC4350807 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761490.2 | XP_015616976.1 | 19 - 20 | 4276 | 119 | |
| GenBank mRNA | AK121517.1 | - | 16 - 17 | 4596 | 119 | |
More Information
Set Membership: |
|

LOC4350807
Genomic Map