Assay Details
| Gene Symbol: LOC4334045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774318.2 | XP_015629804.1 | 7 - 8 | 7054 | 112 | |
| XM_015774319.2 | XP_015629805.1 | 7 - 8 | 7054 | 112 | ||
| XM_015774320.2 | XP_015629806.1 | 7 - 8 | 7054 | 112 | ||
| XM_015774322.2 | XP_015629808.1 | 7 - 8 | 7054 | 112 | ||
More Information
Set Membership: |
|

LOC4334045
Genomic Map