Assay Details
| Gene Symbol: LOC4337476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778021.2 | XP_015633507.1 | 1 - 2 | 169 | 68 | |
| GenBank mRNA | AK061239.1 | - | 1 - 2 | 85 | 68 | |
| AK073929.1 | - | 1 - 2 | 87 | 68 | ||
| AY570726.1 | - | 1 - 2 | 85 | 68 | ||
| BT132283.1 | - | 1 - 2 | 85 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015778021.2 transcripts.
Set Membership: |
|

LOC4337476
Genomic Map