Assay Details
| Gene Symbol: LOC4337476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778021.2 | XP_015633507.1 | 4 - 5 | 391 | 128 | |
| GenBank mRNA | AK061239.1 | - | 4 - 5 | 307 | 128 | |
| AK073929.1 | - | 4 - 5 | 309 | 128 | ||
| AY570726.1 | - | 4 - 5 | 307 | 128 | ||
| BT132283.1 | - | 4 - 5 | 307 | 128 | ||
More Information
Set Membership: |
|

LOC4337476
Genomic Map