Assay Details
| Gene Symbol: LOC4337476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778021.2 | XP_015633507.1 | 5 - 6 | 505 | 87 | |
| GenBank mRNA | AK061239.1 | - | 5 - 6 | 421 | 87 | |
| AK073929.1 | - | 5 - 6 | 423 | 87 | ||
| AY570726.1 | - | 5 - 6 | 421 | 87 | ||
| BT131574.1 | - | 1 - 2 | 112 | 87 | ||
| BT132283.1 | - | 5 - 6 | 421 | 87 | ||
More Information
Set Membership: |
|

LOC4337476
Genomic Map