Assay Details
| Gene Symbol: LOC4337476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778021.2 | XP_015633507.1 | 7 - 8 | 689 | 71 | |
| GenBank mRNA | AK061239.1 | - | 7 - 8 | 605 | 71 | |
| AK073929.1 | - | 7 - 8 | 607 | 71 | ||
| AY570726.1 | - | 7 - 8 | 605 | 71 | ||
| BT131574.1 | - | 3 - 4 | 296 | 71 | ||
| BT132283.1 | - | 7 - 8 | 605 | 71 | ||
More Information
Set Membership: |
|

LOC4337476
Genomic Map