Assay Details
| Gene Symbol: LOC4343955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791566.2 | XP_015647052.1 | 3 - 4 | 359 | 66 | |
| XM_015791567.2 | XP_015647053.1 | 3 - 4 | 359 | 66 | ||
| GenBank mRNA | AK070292.1 | - | 3 - 4 | 347 | 66 | |
More Information
Set Membership: |
|

LOC4343955
Genomic Map