Assay Details
| Gene Symbol: LOC4344584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794258.2 | XP_015649744.1 | 1 - 2 | 188 | 68 | |
| GenBank mRNA | AK065715.1 | - | 1 - 2 | 185 | 68 | |
| AK099222.1 | - | 1 - 2 | 126 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794258.2 transcripts.
Set Membership: |
|

LOC4344584
Genomic Map