Assay Details
| Gene Symbol: LOC4344584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794258.2 | XP_015649744.1 | 2 - 3 | 352 | 61 | |
| GenBank mRNA | AB099292.1 | - | 1 - 2 | 125 | 61 | |
| AK065715.1 | - | 2 - 3 | 349 | 61 | ||
| AK099222.1 | - | 2 - 3 | 290 | 61 | ||
More Information
Set Membership: |
|

LOC4344584
Genomic Map