Assay Details
| Gene Symbol: LOC4344584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794258.2 | XP_015649744.1 | 5 - 6 | 701 | 85 | |
| GenBank mRNA | AB099292.1 | - | 4 - 5 | 474 | 85 | |
| AK065715.1 | - | 5 - 6 | 698 | 85 | ||
| AK099222.1 | - | 5 - 6 | 639 | 85 | ||
More Information
Set Membership: |
|

LOC4344584
Genomic Map