Assay Details
| Gene Symbol: LOC4344584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794258.2 | XP_015649744.1 | 7 - 8 | 931 | 81 | |
| GenBank mRNA | AB099292.1 | - | 6 - 7 | 704 | 81 | |
| AK065715.1 | - | 7 - 8 | 928 | 81 | ||
| AK099222.1 | - | 7 - 8 | 869 | 81 | ||
More Information
Set Membership: |
|

LOC4344584
Genomic Map