Assay Details
| Gene Symbol: LOC4347814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756210.2 | XP_015611696.1 | 12 - 13 | 1817 | 68 | |
| XM_015756211.2 | XP_015611697.1 | 11 - 12 | 1703 | 68 | ||
| GenBank mRNA | AB159444.1 | - | 12 - 13 | 1812 | 68 | |
| AK101142.1 | - | 12 - 13 | 1812 | 68 | ||
More Information
Set Membership: |
|

LOC4347814
Genomic Map