Assay Details
| Gene Symbol: LOC4348262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757392.2 | XP_015612878.1 | 7 - 8 | 992 | 124 | |
| GenBank mRNA | AK099880.1 | - | 3 - 4 | 516 | 124 | |
| AK100514.1 | - | 7 - 8 | 979 | 124 | ||
More Information
Set Membership: |
|

LOC4348262
Genomic Map