Assay Details
Gene Symbol: LOC4352868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764532.1 | XP_015620018.1 | 1 - 2 | 236 | 116 | |
GenBank mRNA | AJ002893.1 | - | 1 - 2 | 199 | 116 | |
AK243636.1 | - | 1 - 2 | 218 | 116 | ||
AK288031.1 | - | 3028 | 116 | |||
AK288365.1 | - | 2965 | 116 | |||
AK289192.1 | - | 218 | 116 | |||
EU267964.1 | - | 1 - 2 | 204 | 116 | ||
GQ848043.1 | - | 1 - 2 | 203 | 116 | ||
JF969238.1 | - | 1 - 2 | 203 | 116 |
Gene Symbol: LOC4326315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK243674.1 | - | 1325 | 116 |
Gene Symbol: LOC4331166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK243152.1 | - | 2293 | 116 |
Gene Symbol: LOC4350992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK243042.1 | - | 2153 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
