Assay Details
| Gene Symbol: LOC4324035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766688.2 | XP_015622174.1 | 3 - 4 | 487 | 121 | |
| XM_015766689.2 | XP_015622175.1 | 2 - 3 | 586 | 121 | ||
| GenBank mRNA | AK111736.1 | - | 3 - 4 | 494 | 121 | |
More Information
Set Membership: |
|

LOC4324035
Genomic Map