Assay Details
Gene Symbol: LOC4325824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015786354.2 | XP_015641840.1 | 5 - 6 | 846 | 71 | |
GenBank mRNA | AK242715.1 | - | 5 - 6 | 822 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
