Assay Details
| Gene Symbol: LOC4333020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775047.2 | XP_015630533.1 | 2 - 3 | 317 | 109 | |
| XM_015775048.2 | XP_015630534.1 | 2 - 3 | 317 | 109 | ||
| XM_015775049.2 | XP_015630535.1 | 2 - 3 | 318 | 109 | ||
| GenBank mRNA | AB110190.1 | - | 2 - 3 | 272 | 109 | |
| AK103600.1 | - | 2 - 3 | 274 | 109 | ||
More Information
Set Membership: |
|

LOC4333020
Genomic Map