Assay Details
| Gene Symbol: LOC4333020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775047.2 | XP_015630533.1 | 3 - 4 | 376 | 59 | |
| XM_015775048.2 | XP_015630534.1 | 3 - 4 | 376 | 59 | ||
| XM_015775049.2 | XP_015630535.1 | 3 - 4 | 377 | 59 | ||
| GenBank mRNA | AB110190.1 | - | 3 - 4 | 331 | 59 | |
| AK103600.1 | - | 3 - 4 | 333 | 59 | ||
More Information
Set Membership: |
|

LOC4333020
Genomic Map