Assay Details
| Gene Symbol: LOC9267158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781696.2 | XP_015637182.1 | 6 - 7 | 2248 | 110 | |
| GenBank mRNA | AK287825.1 | - | 5 - 6 | 1931 | 110 | |
More Information
Set Membership: |
|

LOC9267158
Genomic Map