Assay Details
Gene Symbol: LOC4336158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015779499.1 | XP_015634985.1 | 6 - 7 | 2995 | 63 | |
GenBank mRNA | AK070908.1 | - | 6 - 7 | 2768 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
