Assay Details
| Gene Symbol: LOC4327742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758783.2 | XP_015614269.1 | 12 - 13 | 1368 | 64 | |
| GenBank mRNA | AK062088.1 | - | 3 - 4 | 373 | 64 | |
| AK067201.1 | - | 6 - 7 | 639 | 64 | ||
More Information
Set Membership: |
|

LOC4327742
Genomic Map