Assay Details
Gene Symbol: LOC4327742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758783.2 | XP_015614269.1 | 14 - 15 | 1603 | 100 | |
GenBank mRNA | AK062088.1 | - | 5 - 6 | 608 | 100 | |
AK067201.1 | - | 8 - 9 | 874 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
