Assay Details
Gene Symbol: LOC4324467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767628.2 | XP_015623114.1 | 1 - 2 | 243 | 102 | |
GenBank mRNA | AK073409.1 | - | 1 - 2 | 225 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015767628.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
