Assay Details
Gene Symbol: LOC4324467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767628.2 | XP_015623114.1 | 2 - 3 | 341 | 95 | |
GenBank mRNA | AK073409.1 | - | 2 - 3 | 323 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
