Assay Details
Gene Symbol: LOC4324467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767628.2 | XP_015623114.1 | 5 - 6 | 875 | 72 | |
GenBank mRNA | AK073409.1 | - | 5 - 6 | 857 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
