Assay Details
| Gene Symbol: LOC4349502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759522.2 | XP_015615008.1 | 12 - 13 | 1439 | 80 | |
| GenBank mRNA | AK065123.1 | - | 12 - 13 | 1447 | 80 | |
More Information
Set Membership: |
|

LOC4349502
Genomic Map