Assay Details
Gene Symbol: LOC4350917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761630.2 | XP_015617116.1 | 3 - 4 | 3956 | 75 | |
XM_015761631.2 | XP_015617117.1 | 4 - 5 | 3844 | 75 | ||
XM_015761632.2 | XP_015617118.1 | 4 - 5 | 3884 | 75 | ||
XM_015761633.2 | XP_015617119.1 | 4 - 5 | 3868 | 75 | ||
XM_015761634.2 | XP_015617120.1 | 4 - 5 | 3847 | 75 | ||
XM_015761635.2 | XP_015617121.1 | 4 - 5 | 3824 | 75 | ||
GenBank mRNA | AK062893.1 | - | 1 - 2 | 72 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
