Assay Details
Gene Symbol: LOC4350917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761630.2 | XP_015617116.1 | 6 - 7 | 4199 | 94 | |
XM_015761631.2 | XP_015617117.1 | 7 - 8 | 4087 | 94 | ||
XM_015761632.2 | XP_015617118.1 | 7 - 8 | 4127 | 94 | ||
XM_015761633.2 | XP_015617119.1 | 7 - 8 | 4111 | 94 | ||
XM_015761634.2 | XP_015617120.1 | 7 - 8 | 4090 | 94 | ||
XM_015761635.2 | XP_015617121.1 | 7 - 8 | 4067 | 94 | ||
GenBank mRNA | AK062893.1 | - | 4 - 5 | 315 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
